missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organic Carbon Stock Solution (1000 ppm C) Preserved with Phosphoric Acid, Reagents
Organic Carbon Stock Solution (1000 ppm C) Preserved with Phosphoric Acid, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.
$85.57 - $754.93
Specifications
Chemical Name or Material | Organic Carbon Stock Solution (1000 ppm C) Preserved with Phosphoric Acid |
---|---|
Packaging | Glass Bottle |
CAS | 7732-18-5,877-24-7 |
Specific Gravity | 1.001 |
Physical Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-335-588
|
Labchem Inc
CC205700125C |
125 mL |
Each for $85.57
|
|
|||||
01-335-587
|
Labchem Inc
CC2057001C |
1 L |
Each for $288.63
|
|
|||||
01-335-586
|
Labchem Inc
CC2057004C |
4 L |
Each for $754.93
|
|
|||||
01-335-585
|
Labchem Inc
CC205700500C |
500 mL |
Each for $154.60
|
|
|||||
Specifications
Organic Carbon Stock Solution (1000 ppm C) Preserved with Phosphoric Acid | |
7732-18-5,877-24-7 | |
Liquid | |
<2 | |
Water Analyzer | |
Water Analysis |
Glass Bottle | |
1.001 | |
Colorless | |
100°C | |
Completely miscible | |
Reagent |