missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organic Carbon Stock Solution (1000 ppm C) Preserved with Phosphoric Acid, Reagents
Organic Carbon Stock Solution (1000 ppm C) Preserved with Phosphoric Acid, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CC2057004C
Specifications
Organic Carbon Stock Solution (1000 ppm C) Preserved with Phosphoric Acid | |
7732-18-5,877-24-7 | |
Liquid | |
<2 | |
Water Analyzer | |
Water Analysis | |
Reagent |
Glass Bottle | |
1.001 | |
Colorless | |
100°C | |
Completely miscible | |
4 L |