Learn More
MilliporeSigma™ Supelco™ Omnifit™ EZ Benchmark Columns with 1 Adjustable and 1 Fixed Endpiece
Omnifit™ Labware complete columns are supplied assembled and ready for use, with tubing and fittings to connect the column into an existing system.
Supplier:  MilliporeSigma™ Supelco™ K5114U
 View more versions of this product
Catalog No. 111007939
Only null left
Explore available promotions
Description
Description
This configuration of Benchmark columns have one fixed endpiece as a bed support and one adjustable endpiece (plunger) to adjust the bed height (-AF). The adjustable endpiece is designed to give the user fine control over the movement of the endpiece, minimizing the risk of disturbance to the packed bed and giving 80 mm of height adjustment.
Specifications
Specifications
15 mm
250 mm
Omnifit
Chromatography
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Omnifit is a registered trademark of Diba Industries