missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α-Amylase from Porcine pancreas, MP Biomedicals™
α-Amylase from Porcine pancreas ≥700 u/mg protein, Opaque, White Suspension
$428.27 - $599.30
Specifications
Color | Opaque White |
---|---|
Synonym | β-N-acetylglucosaminidase porcine placenta |
Form | Suspension |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN19123925
|
MP Biomedicals Inc
0219123925 |
25 mg |
Each for $599.30
|
|
|||||
MP219123910
|
MP Biomedicals Inc
0219123910 |
10 mg |
Each for $428.27
|
|
|||||
Description
- It is used to hydrolyze α bonds of α-linked polysaccharides, such as starch and glycogen.
- It has been used in various plant studies, such as metabolism studies in Arabidopsis.
- It is isolated from porcine pancreas is a glycoprotein.
- It is a single polypeptide chain of ∽475 residues containing two SH groups and four disulfide bridges and a tightly bound Ca2+ necessary for stability. Chloride ions are necessary for activity and stability.
Specifications
Opaque White | |
β-N-acetylglucosaminidase porcine placenta | |
Suspension |