missing translation for 'onlineSavingsMsg'
Learn More

MAPA™ RollPruf™ Tripolymer Cleanroom Gloves

Hand-specific, triple polymer gloves provide durability and strength with color band at rolled cuff edge to identify size. Available in pair-pack or bulk-pack quantities.

$575.26 - $630.04

Specifications

Certifications/Compliance 21 CFR 170-199, IEST-RPCC-0052
Certifications/Compliance (Europe) EN ISO 374-5
Cleanroom Class Class 10 (ISO 4)
Color Yellow
Disposable Single-use
View More Specs

Products 5
Catalog Number Mfr. No. Glove Cuff Color Size Packaging Type Price Quantity  
Catalog Number Mfr. No. Glove Cuff Color Size Packaging Type Price Quantity  
11-394-69
View Documents
MAPA™
34513584
Black 7.5 Bulk
Case of 200 Pair for $575.26
Only null left
 
11-394-72
View Documents
MAPA™
34513587
Black 9 Bulk
Case of 200 Pair for $622.72
Only null left
 
19-249-526
View Documents
MAPA™
34513577
Gold 9 Pair-packed
Case of 200 Each for $630.04
Only null left
 
11-394-70
View Documents
MAPA™
34513585
Green 8 Bulk
Case of 200 Pair for $575.26
Only null left
 
19-249-524
View Documents
MAPA™
34513575
Green 8 Pair-packed
Case of 200 Each for $630.04
Only null left
 
Description

Description

  • Made from triple polymer blend of natural latex, neoprene, and nitrile
  • Suitable for Class 10/ISO Class 4 cleanroom applications
  • Recommended for use in semiconductor, electronics, aerospace, pharmaceutical and biotechnology
  • No pigments, fillers, or DOP plasticizers
  • Resists acids, salts, alcohols, ketones, and alkalis
  • Washed with ultrapure deionized water to remove particulate and surface reactive ions
  • Low-level nonvolatile residue protects sensitive products and processes
  • Stretch in palms and across knuckles allows freedom of movement and reduces tension and fatigue
  • Design provides acute sense of touch for precision handling and assembly
  • Color-coded cuff band (colored band does not contain titanium dioxide)
Specifications

Specifications

21 CFR 170-199, IEST-RPCC-0052
Class 10 (ISO 4)
Single-use
No
Powdered
8 mil
RollPruf™
30 cm
EN ISO 374-5
Yellow
Partially Textured
Tripolymer, Neoprene, Nitrile
Non-sterile
0.20 mm
11.8 in.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'