Learn More
Corepoint™ Scientific Manual Defrost Laboratory and Medical Freezer with Hydrocarbon Refrigerant
Offering professional cold storage for scientific and industrial settings.
Supplier:  Corepoint™ Scientific NSPF171WWW0MHC
 View more versions of this product
Catalog No. 361058681
Only null left
Explore available promotions
Description
Description
Units come equipped with:
  • Microprocessor temperature control
  • Temperature alarms
  • Probe access
  • Keyed door locks
  • Natural hydrocarbon refrigerants
  • These versatile freezers provide multi-functional features in a cost-effective design.
  • All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Specifications
Specifications
Laboratory Freezer
481 L
Solid Swing
Scratch-resistant Painted Steel
6
Upright
Microprocessor Temperature Controller
115 V
17 cu. ft.
-15°C to -25°C
1
Painted Steel
cULus, ETL, ENERGYSTAR
NSPF171WWW0MHC
NEMA 5-15
H-drive
Product Suggestions
Product Suggestions