missing translation for 'onlineSavingsMsg'
Learn More

Malic Acid, FCC, 99-100.5%, Spectrum™ Chemical

MA147, 617-48-1, C4H6O5

$336.30 - $5109.27

Chemical Identifiers

CAS 617-48-1
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.09
InChI Key BJEPYKJPYRNKOW-UHFFFAOYNA-N
IUPAC Name 2-hydroxybutanedioic acid
SMILES OC(CC(O)=O)C(O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18604150
View Documents
Spectrum Chemical Mfg Cor
MA147500GM
500 g Poly Bottle
Each for $336.30
Only null left
 
18604151
View Documents
Spectrum Chemical Mfg Cor
MA14725KG
2.5 kg Poly Pail
Each for $1,013.90
Only null left
 
18604152
View Documents
Spectrum Chemical Mfg Cor
MA14712KG
12 kg Poly Pail
Each for $2,917.27
Only null left
 
18604153
View Documents
Spectrum Chemical Mfg Cor
MA14745KGBL
45 kg Fiber Drum
Each for $5,109.27
Only null left
 
Description

Description

Spectrum™ Chemical Malic Acid, FCC is used as a source of extreme tartness. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

617-48-1
134.09
2-hydroxybutanedioic acid
C4H6O5
BJEPYKJPYRNKOW-UHFFFAOYNA-N
OC(CC(O)=O)C(O)=O

Specifications

617-48-1
C4H6O5
OC(CC(O)=O)C(O)=O
134.09
FCC
0.1%
Poly Bottle
100%
BJEPYKJPYRNKOW-UHFFFAOYNA-N
2-hydroxybutanedioic acid
99 to 100.5%
500 g
-0.10° to +0.10°
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'