missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malic Acid, FCC, 99-100.5%, Spectrum™ Chemical
MA147, 617-48-1, C4H6O5
Supplier: Spectrum Chemical Mfg Cor MA14712KG
Description
Spectrum™ Chemical Malic Acid, FCC is used as a source of extreme tartness. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
617-48-1 | |
C4H6O5 | |
OC(CC(O)=O)C(O)=O | |
134.09 | |
FCC | |
0.1% | |
Poly Pail |
100% | |
BJEPYKJPYRNKOW-UHFFFAOYNA-N | |
2-hydroxybutanedioic acid | |
99 to 100.5% | |
12 kg | |
-0.10° to +0.10° |
Chemical Identifiers
617-48-1 | |
134.09 | |
2-hydroxybutanedioic acid |
C4H6O5 | |
BJEPYKJPYRNKOW-UHFFFAOYNA-N | |
OC(CC(O)=O)C(O)=O |