missing translation for 'onlineSavingsMsg'
Learn More

SW Sustainability Solutions KarbonHex™ KX43A Purpose Built Impact-Resistant Gloves with Triple Layer Protection

Triple protection gloves with 75% dipped coating provide liquid, oil, and grease resistance. Nitrile sandy palm for a performance grip. Flexible TPR impact resistance, creating a versatile protection with precision handling.

$882.75 - $882.75

Spécifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) EN388:2016
Chlorinated No
Coating Material Nitrile
Color Black, Gray
voir les spécifications

Produits 5
Numéro de catalogue Numéro du manufacturier. Size Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Size Prix Quantité  
17441167
Afficher les documents
SW Sustainability Solutions
C166772
Small
caisse de 36 chaque for $882.75
Il en reste null
 
17441168
Afficher les documents
SW Sustainability Solutions
C166773
Medium
caisse de 36 chaque for $882.75
Il en reste null
 
17441169
Afficher les documents
SW Sustainability Solutions
C166774
Large
caisse de 36 chaque for $882.75
Il en reste null
 
17441170
Afficher les documents
SW Sustainability Solutions
C166775
X-Large
caisse de 36 chaque for $882.75
Il en reste null
 
17441171
missing translation for 'viewDocuments'
SW Sustainability Solutions
C166776
2X-Large
caisse de 36 chaque for $882.75
Il en reste null
 
Description

Description

  • Integrated expansion slots and flex points provide a low profile dorsal impact protection to fingers and knuckles while remaining flexible and dexterous.
  • Nitrile palm coating with sandy texture provides precision handling in wet and oily conditions.
  • Smooth 3/4 dip nitrile base coating provides excellent liquid, oil and grease resistance.
  • Seamless 15 g knitted nylodex shell provides exceptional fit and all-day comfort.
Spécifications

Spécifications

ANSI 6
No
Black, Gray
Specific
Nylon, Spandex
Mechanical Applications
Elasticated
Impact, Abrasion, Liquid
Straight
25.9 cm
EN388:2016
Nitrile
Fully Textured
Yes
Work Glove
Gray
ANSI/ISEA 105-16 Level 2
C16677
10.2 in.
Videos
FDS
Documents

Documents