missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-4 ProQuantum Immunoassay Kit
ELISA
$990.00 - $4150.00
Spécifications
Accession Number | P05112 |
---|---|
Assay Range | 0.0128 to 5000 pg/mL |
Assay Sensitivity | <0.01 pg/mL |
Conjugate | FAM™ |
Detection Method | Taqman™ Real-time PCR |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
A35587
![]() |
Invitrogen™
A35587 |
96 Tests |
chaque for $990.00
|
|
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
||||
A355875
![]() |
Invitrogen™
A355875 |
5 x 96 Tests |
paquet de 5 chaque for $4,150.00
|
|
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
||||
Description
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.Spécifications
P05112 | |
<0.01 pg/mL | |
Taqman™ Real-time PCR | |
ProQuantum | |
Human | |
qPCR Instruments | |
BCGF-1; BCGF1; BSF-1; BSF1; IL-4 | |
Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
RUO | |
Frozen at -20°C | |
30 mins |
0.0128 to 5000 pg/mL | |
FAM™ | |
Proximity-based Immunoassay | |
Plasma, Serum, Supernatant | |
Protein Quantitation | |
3565 | |
1 hr. | |
FAM™ | |
Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL | |
IL-4 | |
2 hrs |