missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-16 Monoclonal Antibody (K1E006_3F11), Invitrogen™
Mouse Monoclonal Antibody
$1693.48 - $2847.38
Specifications
Antigen | IL-16 |
---|---|
Clone | K1E006_3F11 |
Concentration | 1 mg/mL |
Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Description
Sequence of this protein is as follows: DSAASASAAS DVSVESTAEA TVCTVTLEKM SAGLGFSLEG GKGSLHGDKP LTINRIFKGA ASEQSETVQP GDEILQLGGT AMQGLTRFEA WNIIKALPDG PVTIVIRRKS LQSKETTAAG
IL-16 Monoclonal antibody specifically detects IL-16 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western BlotSpecifications
IL-16 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide | |
FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16 | |
Recombinant human IL16 (immunogen range is 1211-1330). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IL16 |
K1E006_3F11 | |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q14005 | |
3603 | |
IgG2a | |
Protein A | |
Antibody |