missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-16 Monoclonal Antibody (K1E006_3F11), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA555937

 View more versions of this product

Catalog No. MA555937


Only null left
Explore available promotions

Description

Description

Sequence of this protein is as follows: DSAASASAAS DVSVESTAEA TVCTVTLEKM SAGLGFSLEG GKGSLHGDKP LTINRIFKGA ASEQSETVQP GDEILQLGGT AMQGLTRFEA WNIIKALPDG PVTIVIRRKS LQSKETTAAG

The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

IL-16
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
Q14005
IL16
Recombinant human IL16 (immunogen range is 1211-1330).
50 μg
Primary
Human
Antibody
IgG2a
ELISA, Immunohistochemistry (Paraffin), Western Blot
K1E006_3F11
Unconjugated
IL16
FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16
Mouse
Protein A
RUO
3603
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions