missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Hypersil™ MOS C8 LC Column, 5μm
Reliably analyze moderately hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids and environmental samples.
$1550.43 - $1550.43
Specifications
Column Format | Analytical Column |
---|---|
Particle Size | 5 μm |
pH | 2 to 8 |
Pore Size | 120 Å |
Surface Area | 170 m²/g |
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
03-341-999
|
Thermo Scientific™
44513NC |
100 mm | 2.1 mm |
Pack of 500 Each for $1,550.43
|
|
||||
Description
- Reliable, less retentive phase
- Long column lifetimes, even under basic conditions
- Manufactured under ISO 9001-2000 quality guidelines
- Affordable
Specifications
Analytical Column | |
2 to 8 | |
170 m²/g | |
C8 | |
Reversed Phase |
5 μm | |
120 Å | |
L7 | |
6.5% | |
Reversed Phase |