missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, 48%, ACS, 47.0-49.0%, Spectrum™ Chemical

$512.37 - $25556.73

Chemical Identifiers

CAS 10035-10-6
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610841
View Documents
Spectrum Chemical Mfg Cor
H1030120ML
120 mL Amber Glass Bottle
Each for $512.37
Only null left
 
18610849
View Documents
Spectrum Chemical Mfg Cor
H1030500MLGL
500 mL Amber Glass Bottle
Each for $840.27
Only null left
 
18610848
View Documents
Spectrum Chemical Mfg Cor
H103025LTPV
2.5 L Poly-Coated Glass Bottle
Each for $2,891.73
Only null left
 
18610840
View Documents
Spectrum Chemical Mfg Cor
H10304LTGL
4 L Amber Glass Bottle
Each for $3,170.93
Only null left
 
18610843
View Documents
Spectrum Chemical Mfg Cor
H103020LTBL
20 L Poly Drum
Each for $6,298.13
Only null left
 
18610842
View Documents
Spectrum Chemical Mfg Cor
H1030200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

10035-10-6

Specifications

10035-10-6
Amber Glass Bottle
120 mL
ACS
5 ppm
2e-05
47 to 49%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.