missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, 48%, ACS, 47.0-49.0%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor H103025LTPV

 View more versions of this product

Catalog No. 18610848


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10035-10-6
Poly-Coated Glass Bottle
2.5 L
ACS
5 ppm
2e-05
47 to 49%

Chemical Identifiers

10035-10-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.