Learn More
MilliporeSigma™ Supelco™ Hamilton™ Needles for RN Syringes, Bevel Tip, Stainless Steel
Hamilton™ needles for RN syringes are bevel tip needles, designed for optimum septum penetration and to prevent septum coring.
$77.41 - $594.72
Specifications
Material Stainless Steel (Tip)
Product Type Bevel Tip
Quantity 6/Pk.
For Use With (Equipment) Hamilton RN Syringes
View More Specs
Products 7
Catalog Number Mfr. No. Needle Gauge Volume (Metric) Length (Metric) Needle Price Quantity  
Catalog Number Mfr. No. Needle Gauge Volume (Metric) Length (Metric) Needle Price Quantity  
111011620
View Documents
MilliporeSigma™ Supelco™
Z2889931PAK
22 G 2.5 to 100 μL 51 mm
Each for $77.41
Only null left
111011622
View Documents
MilliporeSigma™ Supelco™
19133U
22s G 2.5 to 100 μL 51 mm
Each for $77.41
Only null left
111011621
View Documents
MilliporeSigma™ Supelco™
19131U
26s G 2.5 to 100 μL 51 mm
Each for $78.64
Only null left
111010862
View Documents
MilliporeSigma™ Supelco™
Z1181176EA
22 G 2.5 to 100 μL 76 mm
Each for $524.30
Only null left
111011623
View Documents
MilliporeSigma™ Supelco™
19137U
22 G 250 μL to 10 mL 51 mm
Each for $77.41
Only null left
111011564
View Documents
MilliporeSigma™ Supelco™
19136U
22s G 250 μL to 10 mL 51 mm
Each for $81.80
Only null left
111010754
View Documents
MilliporeSigma™ Supelco™
Z1181096EA
22 G 250 μL to 10 mL 101.5 mm
Each for $594.72
Only null left
Description
Description
Specifications
Specifications
Stainless Steel (Tip)
Bevel Tip
6/Pk.
Hamilton RN Syringes
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Hamilton is a registered trademark of Hamilton Co.