Learn More
MilliporeSigma™ Supelco™ Hamilton™ Needles for RN Syringes, Bevel Tip, Stainless Steel
Hamilton™ needles for RN syringes are bevel tip needles, designed for optimum septum penetration and to prevent septum coring.
Supplier: MilliporeSigma™ Supelco™ 19131U
Specifications
Stainless Steel (Tip) | |
2 in. | |
6/Pk. | |
51 mm | |
Hamilton RN Syringes |
26s G | |
Bevel Tip | |
2.5 to 100 μL | |
2 |
Hamilton is a registered trademark of Hamilton Co.