Learn More
MilliporeSigma™ Supelco™ Hamilton™ Needles for LT and TLL Syringes, Metal Hub (N)
Supplier:  MilliporeSigma™ Supelco™ 20803
 View more versions of this product
Catalog No. 111010467
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Stainless Steel (Tip)
Metal Hub (N)
Cone Tip
51 mm
Hamilton LT and TLL Syringes
22 G
2 in.
6/Pk.
5
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Hamilton is a registered trademark of Hamilton Co.