missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 96%, Reagent, ACS, 96%, Spectrum™ Chemical

F1089, 64-18-6, CH2O2

Supplier:  Spectrum Chemical Mfg Cor F108925LTPVCS4

 View more versions of this product

Catalog No. 18602843


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Formic Acid, 96 Percent, Reagent, ACS , also known as methanoic acid, occurs naturally in ant venom. One of its biggest uses is as a preservative agent in livestock feed. It is equally used in the production of leather goods. In the lab it is used as a source of formyl or hydride ion. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Specifications

Specifications

64-18-6
CH2O2
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
96%
4 x 2.5 L
0.003%
100%
UN1779
OC=O
46.03
ACS
Poly-Coated Glass Bottle

Chemical Identifiers

64-18-6
46.03
formic acid
CH2O2
BDAGIHXWWSANSR-UHFFFAOYSA-N
OC=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions