missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid, 96%, Reagent, ACS, 96%, Spectrum™ Chemical
F1089, 64-18-6, CH2O2
Supplier: Spectrum Chemical Mfg Cor F108925LTPV
Description
Spectrum™ Chemical Formic Acid, 96 Percent, Reagent, ACS , also known as methanoic acid, occurs naturally in ant venom. One of its biggest uses is as a preservative agent in livestock feed. It is equally used in the production of leather goods. In the lab it is used as a source of formyl or hydride ion. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.Specifications
64-18-6 | |
CH2O2 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
96% | |
2.5 L | |
0.003% |
100% | |
UN1779 | |
OC=O | |
46.03 | |
ACS | |
Poly-Coated Glass Bottle |
Chemical Identifiers
64-18-6 | |
46.03 | |
formic acid |
CH2O2 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
OC=O |