Learn More
Innovatek EZTest Steam G. stearothermophilus
Innovatek EZTest - Steam G.stearothermophilus is an easy-to-use test for monitoring sterilization
Supplier: Innovatek 4140EZS6
Description
- EZTest is a self-contained biological indicator (BI) for monitoring sterilization
- Bacterial spores on a carrier material packaged within a small, thermoplastic culture tube
- Sealed-glass ampoule of specially formulated soybean casein digest culture medium
- Color indicator turns noticeably yellow when growth occurs
- Easy-to-use in-office; no laboratory transfer needed
- Dramatic visual results within 24 hours (48 hours for ethylene oxide)
Specifications
Yellow Indicator | |
EZTest™ | |
24 hours at 57°C | |
106 |
Monitoring Sterilization | |
57°C | |
Soybean Casein Digest Culture Medium, Color Indicator | |
100 Vials/Box |