missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol, Reagent, 99%, Spectrum™ Chemical
E1051, 107-21-1, C2H6O2
$218.73 - $12938.20
Chemical Identifiers
CAS | 107-21-1 |
---|---|
Molecular Formula | C2H6O2 |
Molecular Weight (g/mol) | 62.07 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
IUPAC Name | ethane-1,2-diol |
SMILES | OCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18602559
|
Spectrum Chemical Mfg Cor
E1051500MLPL |
500 mL | Poly Bottle |
Each for $218.73
|
|
||||
18602561
|
Spectrum Chemical Mfg Cor
E10511LTPL |
1 L | Poly Bottle |
Each for $369.27
|
|
||||
18602560
|
Spectrum Chemical Mfg Cor
E1051500MLPLCS6 |
6 x 500 mL | Poly Bottle |
Case of 6 Each for $1,017.90
|
|
||||
18602562
|
Spectrum Chemical Mfg Cor
E10514LTGL |
4 L | Amber Glass Bottle |
Each for $740.27
|
|
||||
18602563
|
Spectrum Chemical Mfg Cor
E10514LTPL |
4 L | Poly Bottle |
Each for $675.20
|
|
||||
18602564
|
Spectrum Chemical Mfg Cor
E10511LTPLCS6 |
6 x 1 L | Poly Bottle |
Case of 6 Each for $1,737.77
|
|
||||
18602565
|
Spectrum Chemical Mfg Cor
E10514LTGLCS4 |
4 x 4 L | Amber Glass Bottle |
Case of 4 Each for $2,477.57
|
|
||||
18602566
|
Spectrum Chemical Mfg Cor
E10514LTPLCS4 |
4 x 4 L | Poly Bottle |
Case of 4 Each for $2,344.90
|
|
||||
18602567
|
Spectrum Chemical Mfg Cor
E105120LTPL |
20 L | Poly Drum |
Each for $3,748.83
|
|
||||
18602568
|
Spectrum Chemical Mfg Cor
E105155GLBL |
55 gal. | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Ethylene Glycol, Reagent is a colorless, odorless, sweet tasting organic compound that has a syrupy texture and is a liquid. It has many lab applications including use as an antifreeze, use as a desiccant, protecting group for carbonyls during chemical synthesis and as a preservative for biological specimens. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.Chemical Identifiers
107-21-1 | |
62.07 | |
ethane-1,2-diol |
C2H6O2 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
OCCO |
Specifications
107-21-1 | |
0.005% | |
Poly Bottle | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
99% |
100% | |
500 mL | |
C2H6O2 | |
OCCO | |
62.07 | |
Reagent |