missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, Reagent, 99%, Spectrum™ Chemical

E1051, 107-21-1, C2H6O2

Supplier:  Spectrum Chemical Mfg Cor E10514LTGL

 View more versions of this product

Catalog No. 18602562


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Ethylene Glycol, Reagent is a colorless, odorless, sweet tasting organic compound that has a syrupy texture and is a liquid. It has many lab applications including use as an antifreeze, use as a desiccant, protecting group for carbonyls during chemical synthesis and as a preservative for biological specimens. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Specifications

Specifications

107-21-1
0.005%
Amber Glass Bottle
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
99%
100%
4 L
C2H6O2
OCCO
62.07
Reagent

Chemical Identifiers

107-21-1
62.07
ethane-1,2-diol
C2H6O2
LYCAIKOWRPUZTN-UHFFFAOYSA-N
OCCO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions