missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 60 NexGen™ Boot Covers

Boot covers with PVC skid-resistant soles are suitable for protection against non-hazardous particles, light liquid splash, and aerosols in many industrial applications.

$267.81 - $290.45

Specifications

Material Polypropylene
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-715-314
View Documents
DuPont™
NG456SWHMD001000
Medium
Case of 100 Each for $290.45
Only null left
 
19-715-315
View Documents
DuPont™
NG456SWHLG001000
Large
Case of 100 Each for $267.81
Only null left
 
Description

Description

  • Made with a microporous film laminated to a spundbonded polypropylene substrate
  • Applications include automotive refinishing, waste cleanup, and remediation
  • Boot cover height is 18 in.
  • Elastic top provides coverage up to calf
  • PVC soles provides enhanced skid-resistance and durability
  • One size fits most
Specifications

Specifications

Polypropylene
Single-use
Elastic Top
Serged
Automotive, Refinishing, Waste Cleanup, Remediation
45.72 cm
Boot Covers
White
Non-sterile
Non-skid
10 mil
18 in.
Dry Particulates, Fluid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'