missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 60 NexGen™ Boot Covers
Boot covers with PVC skid-resistant soles are suitable for protection against non-hazardous particles, light liquid splash, and aerosols in many industrial applications.
Supplier: DuPont™ NG456SWHLG001000
Description
- Made with a microporous film laminated to a spundbonded polypropylene substrate
- Applications include automotive refinishing, waste cleanup, and remediation
- Boot cover height is 18 in.
- Elastic top provides coverage up to calf
- PVC soles provides enhanced skid-resistance and durability
- One size fits most
Specifications
Large | |
White | |
Non-sterile | |
Non-skid | |
Serged | |
Automotive, Refinishing, Waste Cleanup, Remediation | |
35.56 cm | |
45.72 cm | |
Boot Covers |
Polypropylene | |
Single-use | |
Elastic Top | |
NG456SWHLG010000 | |
10 mil | |
14 in. | |
18 in. | |
Dry Particulates, Fluid |