Learn More
Beckman Coulter Dry Powder System (DPS) for LS 13 320 XR Particle Size Analyzer
A sampling module for the LS 13 320 XR Particle Size Analyzer. The DPS is capable of feeding and measuring dry powder samples in the size range 0.40 μm to 3500 μm. The sample is placed in a DPS tube and delivered to the sensing zone in the optical bench by a required vacuum.
Supplier:  Beckman Coulter B98103
Catalog No. 15655166
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Measures entire sample as required by the ISO 13 320 Standard
  • Programmable Obscuration setting to optimize sample feed rate
  • User-selectable vacuum pressure for maximum dispersion control
  • Sample is placed in a sample holder and delivered to the sensing zone in the optical bench by a vacuum
  • Requires HiCap HEPA Vacuum or Standard LS Vacuum
  • Analytical size range: 0.40 μm to 3,500 μm
Specifications
Specifications
Sampling Module
0.40 μm to 3500 μm
LS 13 320 XR Particle Size Analyzer
Dry Powder System (DPS)
Product Suggestions
Product Suggestions