missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dithieno[3,2-b:2',3'-d]thiophene 98.0+%, TCI America™
$175.22 - $596.49
Chemical Identifiers
CAS | 3593-75-7 |
---|---|
Molecular Formula | C8H4S3 |
Molecular Weight (g/mol) | 196.30 |
MDL Number | MFCD00506977 |
InChI Key | VGWBXRXNERKBSJ-UHFFFAOYSA-N |
PubChem CID | 137985 |
IUPAC Name | 3,7,11-trithiatricyclo[6.3.0.0²,â¶]undeca-1(8),2(6),4,9-tetraene |
SMILES | S1C=CC2=C1C1=C(S2)C=CS1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
D3641200MG
|
TCI America
D3641200MG |
200 mg |
Each for $175.22
|
|
|||||
D36411G
|
TCI America
D36411G |
1 g |
Each for $596.49
|
|
|||||
Chemical Identifiers
3593-75-7 | |
196.30 | |
VGWBXRXNERKBSJ-UHFFFAOYSA-N | |
3,7,11-trithiatricyclo[6.3.0.0²,â¶]undeca-1(8),2(6),4,9-tetraene |
C8H4S3 | |
MFCD00506977 | |
137985 | |
S1C=CC2=C1C1=C(S2)C=CS1 |
Specifications
3593-75-7 | |
138°C | |
MFCD00506977 | |
VGWBXRXNERKBSJ-UHFFFAOYSA-N | |
3,7,11-trithiatricyclo[6.3.0.0²,â¶]undeca-1(8),2(6),4,9-tetraene | |
137985 | |
≥98.0% (GC) | |
Dithieno[3,2-b:2′,3′-d]thiophene |
67°C | |
C8H4S3 | |
200 mg | |
S1C=CC2=C1C1=C(S2)C=CS1 | |
196.30 | |
196.30 | |
Crystalline Powder |
Safety and Handling
TSCA : No