missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dithieno[3,2-b:2',3'-d]thiophene 98.0+%, TCI America™
Supplier: TCI America D36411G
Specifications
Dithieno[3,2-b:2′,3′-d]thiophene | |
67°C | |
C8H4S3 | |
1 g | |
S1C=CC2=C1C1=C(S2)C=CS1 | |
196.30 | |
196.30 | |
Crystalline Powder |
3593-75-7 | |
138°C | |
MFCD00506977 | |
VGWBXRXNERKBSJ-UHFFFAOYSA-N | |
3,7,11-trithiatricyclo[6.3.0.0²,â¶]undeca-1(8),2(6),4,9-tetraene | |
137985 | |
≥98.0% (GC) |
Chemical Identifiers
3593-75-7 | |
196.30 | |
VGWBXRXNERKBSJ-UHFFFAOYSA-N | |
3,7,11-trithiatricyclo[6.3.0.0²,â¶]undeca-1(8),2(6),4,9-tetraene |
C8H4S3 | |
MFCD00506977 | |
137985 | |
S1C=CC2=C1C1=C(S2)C=CS1 |
Safety and Handling
TSCA : No