missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Discovery™ C18 HPLC Column, 5 μm Particle Size
Use Discovery™ C18 for any method that specifies a C18. The exceptional peak shape, reproducibility, and stability make it the column of choice for all C18 methods from demanding to routine.
Supplier: MilliporeSigma™ Supelco™ 50497130
Description
- Excellent reproducibility
- Exceptional peak shape for basic and acidic analytes
- Stable, low-bleed LC-MS separations
- Separation of peptides and small proteins
- Lower hydrophobicity than many comparable C18 columns, providing faster analysis
Specifications
HPLC, LC/MS Column | |
25 cm | |
5 μm | |
180 Å | |
≤70°C | |
C18 (Octadecyl) | |
C18 (Octadecyl) Phase/Silica Gel, High Purity, Spherical Particle Platform | |
12% | |
Yes | |
Stainless Steel | |
LC/MS |
3 mm | |
400 bar(5801 psi) | |
2 to 8 | |
200 m2/g | |
USP L1 | |
Discovery | |
Silica gel, high purity, spherical particle platform fully porous particle | |
Reversed Phase | |
Reversed Phase | |
LC-MS Column |
Discovery is a registered trademark of Sigma-Aldrich Co. LLC