missing translation for 'onlineSavingsMsg'
Learn More

Butyl Butyrate, 98%, Spectrum™ Chemical

$201.29 - $18674.47

Chemical Identifiers

CAS 109-21-7
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609380
View Documents
Spectrum Chemical Mfg Cor
B1183500ML
500 mL Amber Glass Bottle
Each for $201.29
Only null left
 
18609379
View Documents
Spectrum Chemical Mfg Cor
B11834LTGL
4 L Amber Glass Bottle
Each for $847.00
Only null left
 
18609377
View Documents
Spectrum Chemical Mfg Cor
B118320LT
20 L Steel Can
Each for $3,145.76
Only null left
 
18609378
View Documents
Spectrum Chemical Mfg Cor
B1183200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

109-21-7

Specifications

1
1
1.405-1.407
0.867 - 0.871
Reagent
109-21-7
Amber Glass Bottle
500 mL
0.98
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.