missing translation for 'onlineSavingsMsg'
Learn More

Butyl Butyrate, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor B118320LT

 View more versions of this product

Catalog No. 18609377


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1
1
1.405-1.407
0.867 - 0.871
Reagent
109-21-7
Steel Can
20 L
0.98

Chemical Identifiers

109-21-7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.