missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Bottles, Storage, Wide Mouth
Safely store liquids and powders.
Supplier: United Scientific™ 81900
Description
- These bottles provide an excellent option for storage of reagents and other materials in the laboratory.
- The square shape is designed to optimize shelf space.
- Available in three different sizes, these bottles feature polypropylene caps, are autoclavable, and provide good chemical resistance.
Specifications
250 mL | |
Clear | |
12/Pk. | |
Yes | |
Reusable | |
Threaded | |
87 mm | |
Lab Scoops | |
36 mm ID | |
No |
8.45 oz. | |
Polypropylene | |
Non-sterile | |
Autoclavable | |
No | |
Square | |
Square Wide Mouth Bottle | |
Polypropylene | |
Screw Cap |