missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Bottles, Storage, Wide Mouth
Safely store liquids and powders.
$56.17 - $81.63
Specifications
Color | Clear |
---|---|
Material | Polypropylene |
Sterility | Non-sterile |
Closure Included | Yes |
Autoclavable | Autoclavable |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
S15744
![]() |
United Scientific™
81901 |
500 mL |
Pack of 12 Each for $81.63
|
|
|||||
S15757
![]() |
United Scientific™
81900 |
250 mL |
Pack of 12 Each for $67.40
|
|
|||||
S15940
![]() |
United Scientific™
81902 |
1000 mL |
Pack of 6 Each for $56.17
|
|
|||||
Description
- These bottles provide an excellent option for storage of reagents and other materials in the laboratory.
- The square shape is designed to optimize shelf space.
- Available in three different sizes, these bottles feature polypropylene caps, are autoclavable, and provide good chemical resistance.
Specifications
Clear | |
Non-sterile | |
Autoclavable | |
No | |
Square | |
Lab Scoops | |
Screw Cap |
Polypropylene | |
Yes | |
Reusable | |
Threaded | |
Square Wide Mouth Bottle | |
Polypropylene | |
No |