Learn More
MilliporeSigma BIOshell A160 Peptide PCS-C18 HPLC Column
BIOshell™ A160 Peptide PCS-C18 columns are specifically engineered to provide efficient separation of peptides as well as small proteins.
$1137.48 - $1856.25
Specifications
Particle Size | 2.7 μm |
---|---|
Pore Size | 160 Å |
Phase | Positively Charged Surface (PCS) |
Material | Stainless Steel |
Type | HPLC Column |
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity | ||||
01-000-908
|
MilliporeSigma
50680U |
4.6 mm | 5 mm |
Each for $1,137.48
|
|
||||
01-000-909
|
MilliporeSigma
50675U |
3.0 mm | 5 mm |
Each for $1,137.48
|
|
||||
01-000-910
|
MilliporeSigma
50671U |
2.1 mm | 5 mm |
Each for $1,137.48
|
|
||||
01-000-911
|
MilliporeSigma
50668U |
4.6 mm | 150 mm |
Each for $1,814.38
|
|
||||
01-000-912
|
MilliporeSigma
50665U |
4.6 mm | 100 mm |
Each for $1,660.86
|
|
||||
01-000-913
|
MilliporeSigma
50664U |
4.6 mm | 50 mm |
Each for $1,479.42
|
|
||||
01-000-914
|
MilliporeSigma
50663U |
3.0 mm | 150 mm |
Each for $1,814.38
|
|
||||
01-000-915
|
MilliporeSigma
50661U |
3.0 mm | 100 mm |
Each for $1,660.86
|
|
||||
01-000-916
|
MilliporeSigma
50660U |
3.0 mm | 50 mm |
Each for $1,479.42
|
|
||||
01-000-917
|
MilliporeSigma
50659U |
2.1 mm | 150 mm |
Each for $1,814.38
|
|
||||
01-000-918
|
MilliporeSigma
50657U |
2.1 mm | 100 mm |
Each for $1,660.86
|
|
||||
01-000-919
|
MilliporeSigma
50656U |
2.1 mm | 50 mm |
Each for $1,479.42
|
|
||||
01-000-920
|
MilliporeSigma
50655U |
1.5 mm | 150 mm |
Each for $1,856.25
|
|
||||
01-000-921
|
MilliporeSigma
50654U |
1.5 mm | 100 mm |
Each for $1,702.72
|
|
||||
01-000-922
|
MilliporeSigma
50653U |
1.5 mm | 50 mm |
Each for $1,535.25
|
|
||||
Description
These columns contain advanced Fused-Core™ particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping.
- The unique PCS-C18 chemistry enables exceptional peak shape and enhanced loading capacity of basic peptides as compared to traditional C18 chemistries.
- In addition, the PCS-C18 facilitates the use of mobile phases with lower concentrations of mobile phase modifiers without any compromise to column performance.
- This column also provides orthogonal selectivity to ES-C18, ES-CN (cyano), and phenyl-hexyl chemistries.
Specifications
2.7 μm | |
Positively Charged Surface (PCS) | |
HPLC Column |
160 Å | |
Stainless Steel | |
HPLC |