missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium Chloride, USP, 97-103%, Spectrum™ Chemical

BE129, 121-54-0, C27H42ClNO2

Chemical Identifiers

CAS 121-54-0
Molecular Formula C27H42ClNO2
Molecular Weight (g/mol) 448.09
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
IUPAC Name benzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
SMILES [Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600886
View Documents
Spectrum Chemical Mfg Cor
BE12912KGBL
12 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Benzethonium Chloride, USP exhibits a broad spectrum of microbiocidal activity against bacteria, fungi, mold and viruses. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

121-54-0
448.09
benzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
C27H42ClNO2
UREZNYTWGJKWBI-UHFFFAOYSA-M
[Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1

Specifications

121-54-0
0.1%
Amber Glass Bottle
12 kg
UREZNYTWGJKWBI-UHFFFAOYSA-M
benzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
97 to 103%
Crystalline Powder or Lumps
100%
5%
C27H42ClNO2
UN3261
[Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1
448.09
USP
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'