missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAY 11-7085 97.0+%, TCI America™
$153.06 - $910.87
Chemical Identifiers
CAS | 196309-76-9 |
---|---|
Molecular Formula | C13H15NO2S |
Molecular Weight (g/mol) | 249.328 |
MDL Number | MFCD01862602 |
InChI Key | VHKZGNPOHPFPER-ONNFQVAWSA-N |
Synonym | e-3-4-tert-butylphenylsulfonyl acrylonitrile, e-3-4-tert-butyl phenyl sulfonyl acrylonitrile, e-3-4-t-butylphenylsulfonyl-2-propenenitrile, e-3-4-tert-butylphenyl sulfonylprop-2-enenitrile, e 3-4-t-butylphenyl sulfonyl-2-propenenitrile, 2-propenenitrile, 3-4-1,1-dimethylethyl phenyl sulfonyl-, 2e, 2e-3-4-tert-butylbenzenesulfonyl prop-2-enenitrile, bay hplc , solid, e-3-4-tert-butyl-benzenesulfonyl-acrylonitrile, e-3-4-tert-butylphenyl sulfonyl-2-propenenitrile |
PubChem CID | 5353432 |
IUPAC Name | (E)-3-(4-tert-butylphenyl)sulfonylprop-2-enenitrile |
SMILES | CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)C=CC#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B484510MG
|
TCI America
B484510MG |
10 mg |
Each for $153.06
|
|
|||||
B4845100MG
|
TCI America
B4845100MG |
100 mg |
Each for $910.87
|
|
|||||
Chemical Identifiers
196309-76-9 | |
249.328 | |
VHKZGNPOHPFPER-ONNFQVAWSA-N | |
5353432 | |
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)C=CC#N |
C13H15NO2S | |
MFCD01862602 | |
e-3-4-tert-butylphenylsulfonyl acrylonitrile, e-3-4-tert-butyl phenyl sulfonyl acrylonitrile, e-3-4-t-butylphenylsulfonyl-2-propenenitrile, e-3-4-tert-butylphenyl sulfonylprop-2-enenitrile, e 3-4-t-butylphenyl sulfonyl-2-propenenitrile, 2-propenenitrile, 3-4-1,1-dimethylethyl phenyl sulfonyl-, 2e, 2e-3-4-tert-butylbenzenesulfonyl prop-2-enenitrile, bay hplc , solid, e-3-4-tert-butyl-benzenesulfonyl-acrylonitrile, e-3-4-tert-butylphenyl sulfonyl-2-propenenitrile | |
(E)-3-(4-tert-butylphenyl)sulfonylprop-2-enenitrile |
Specifications
196309-76-9 | |
White | |
MFCD01862602 | |
e-3-4-tert-butylphenylsulfonyl acrylonitrile, e-3-4-tert-butyl phenyl sulfonyl acrylonitrile, e-3-4-t-butylphenylsulfonyl-2-propenenitrile, e-3-4-tert-butylphenyl sulfonylprop-2-enenitrile, e 3-4-t-butylphenyl sulfonyl-2-propenenitrile, 2-propenenitrile, 3-4-1,1-dimethylethyl phenyl sulfonyl-, 2e, 2e-3-4-tert-butylbenzenesulfonyl prop-2-enenitrile, bay hplc , solid, e-3-4-tert-butyl-benzenesulfonyl-acrylonitrile, e-3-4-tert-butylphenyl sulfonyl-2-propenenitrile | |
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)C=CC#N | |
249.328 | |
249.33 | |
Crystalline Powder |
84°C | |
C13H15NO2S | |
10 mg | |
VHKZGNPOHPFPER-ONNFQVAWSA-N | |
(E)-3-(4-tert-butylphenyl)sulfonylprop-2-enenitrile | |
5353432 | |
≥97.0% (N) | |
BAY 11-7085 |
Safety and Handling
RTECSNumber : UD1312500
TSCA : No
Recommended Storage : Refrigerator