missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAY 11-7085 97.0+%, TCI America™
Supplier: TCI America B4845100MG
Specifications
BAY 11-7085 | |
84°C | |
C13H15NO2S | |
100 mg | |
VHKZGNPOHPFPER-ONNFQVAWSA-N | |
(E)-3-(4-tert-butylphenyl)sulfonylprop-2-enenitrile | |
5353432 | |
≥97.0% (N) |
196309-76-9 | |
White | |
MFCD01862602 | |
e-3-4-tert-butylphenylsulfonyl acrylonitrile, e-3-4-tert-butyl phenyl sulfonyl acrylonitrile, e-3-4-t-butylphenylsulfonyl-2-propenenitrile, e-3-4-tert-butylphenyl sulfonylprop-2-enenitrile, e 3-4-t-butylphenyl sulfonyl-2-propenenitrile, 2-propenenitrile, 3-4-1,1-dimethylethyl phenyl sulfonyl-, 2e, 2e-3-4-tert-butylbenzenesulfonyl prop-2-enenitrile, bay hplc , solid, e-3-4-tert-butyl-benzenesulfonyl-acrylonitrile, e-3-4-tert-butylphenyl sulfonyl-2-propenenitrile | |
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)C=CC#N | |
249.328 | |
249.33 | |
Crystalline Powder |
Chemical Identifiers
196309-76-9 | |
249.328 | |
VHKZGNPOHPFPER-ONNFQVAWSA-N | |
5353432 | |
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)C=CC#N |
C13H15NO2S | |
MFCD01862602 | |
e-3-4-tert-butylphenylsulfonyl acrylonitrile, e-3-4-tert-butyl phenyl sulfonyl acrylonitrile, e-3-4-t-butylphenylsulfonyl-2-propenenitrile, e-3-4-tert-butylphenyl sulfonylprop-2-enenitrile, e 3-4-t-butylphenyl sulfonyl-2-propenenitrile, 2-propenenitrile, 3-4-1,1-dimethylethyl phenyl sulfonyl-, 2e, 2e-3-4-tert-butylbenzenesulfonyl prop-2-enenitrile, bay hplc , solid, e-3-4-tert-butyl-benzenesulfonyl-acrylonitrile, e-3-4-tert-butylphenyl sulfonyl-2-propenenitrile | |
(E)-3-(4-tert-butylphenyl)sulfonylprop-2-enenitrile |
Safety and Handling
RTECSNumber : UD1312500
TSCA : No
Recommended Storage : Refrigerator