missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ AP800 ESD Nitrile-Coated Gloves

Electrostatic dissipative 13-gauge micro fiber polyester, embossed microporous foam nitrile coating glove

$808.91 - $808.91

Specifications

Accelerator Free Yes
Applications Industrial
Coating Material Nitrile Foam
Color Gray
Disposable Reusable
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
18-139-747
View Documents
SHOWA™
AP800L08
Small
Case of 144 Pair for $808.91
Only null left
 
18-139-746
View Documents
SHOWA™
AP800M07
Medium
Dozen of 12 Pair for $808.91
Only null left
 
18-139-745
View Documents
SHOWA™
AP800S06
Large
Dozen of 12 Pair for $808.91
Only null left
 
18-139-748
View Documents
SHOWA™
AP800XL09
X-Large
Dozen of 12 Pair for $808.91
Only null left
 
18-139-749
View Documents
SHOWA™
AP800XXL10
2X-Large
Dozen of 12 Pair for $808.91
Only null left
 
Description

Description

The new SHOWA AP800 has a 13-gauge seamless knit with a Microfiber polyester static dissipative yarn and an embossed microporous foam nitrile palm dip. It is free of natural rubber latex and is touchscreen compatible, silicone-free and fingerprint-free. Certified under the standard EN 16350, the AP800 by SHOWA meets the requirements for electrostatic properties (ESD). This certification ensures the glove’s safe usage in ATEX zones (working area with risk of explosion and/or high flammability).
Specifications

Specifications

Yes
Nitrile Foam
Reusable
Ambidextrous
Polyester
ESD Gloves
13 gauge
Industrial
Gray
No Texture
Yes
Powder Free
Non-sterile
0.7 mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'