missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Certified, 0.200N ±0.005N (0.2M), LabChem™

$62.77 - $132.57

Identifiants chimiques

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
LC103602
Afficher les documents
LabChem
LC103602
1 L
chaque for $62.77
Il en reste null
 
LC103604
Afficher les documents
LabChem
LC103604
4 L
chaque for $132.57
Il en reste null
 
Description

Description

Spécifications

Identifiants chimiques

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Spécifications

64-19-7,7732-18-5
Colorless
CH3COOH
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
60.05
CHEBI:15366
Certified
1 L
Poly Bottle
1g/mL
Acetic Acid
98.8,1.2
C2H4O2
MFCD00036152
Soluble in water
CC(O)=O
0.200N ±0.005N (0.2M)
176
60.05
1g/mL
Vinegar
Liquid
Traceable to NIST
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes skin and serious eye irritation.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature

FDS
missing translation for 'documents'

missing translation for 'documents'