Learn More
Corning™ 3D Clear Tissue Clearing Reagent
Corning™'s 3D Clear tissue clearing technique is designed specifically to support imaging for 3D cell culture models and plate-based high-throughput processing. When paired with fluorescent labeling (e.g. fluorescent protein, immunofluorescence, chemical dyes) and high content confocal microscopy, Corning™'s 3D Clear Reagent allows for complete 3D cell culture model characterization and more accurate drug screening.
Supplier:  Corning™ 5731
 View more versions of this product
Catalog No. 10-320-104
Only null left
Explore available promotions
Description
Description
  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent labels
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
Specifications
Specifications
10 mL
Product Suggestions
Product Suggestions