Learn More
Corning™ 3D Clear Tissue Clearing Reagent
Corning™'s 3D Clear tissue clearing technique is designed specifically to support imaging for 3D cell culture models and plate-based high-throughput processing. When paired with fluorescent labeling (e.g. fluorescent protein, immunofluorescence, chemical dyes) and high content confocal microscopy, Corning™'s 3D Clear Reagent allows for complete 3D cell culture model characterization and more accurate drug screening.
$1120.99 - $1120.99
Specifications
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
10-320-105
|
Corning™
5732 |
30 mL |
Case for $1,120.99
|
|
|||||
Description
- Rapid tissue clearing
- Easy to Use
- No Special Equipment Required
- Compatible with IF, FP and other Fluorescent labels
- Microplate and Automation Compatible
- Reversible to Follow up with 2D H&E/IHC
Specifications
0.0 |