missing translation for 'onlineSavingsMsg'
Learn More

Zinc Gluconate, USP, 97-102%, Spectrum™ Chemical

Z1077, 4468-02-4, C12H22O14Zn

$338.29 - $7782.14

Chemical Identifiers

CAS 4468-02-4
Molecular Formula C12H22O14Zn
Molecular Weight (g/mol) 455.67
MDL Number MFCD00868110
InChI Key WHMDKBIGKVEYHS-IYEMJOQQSA-L
IUPAC Name zinc(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
SMILES [Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18607850
View Documents
Spectrum Chemical Mfg Cor
Z1077500GM
500 g Poly Bottle
Each for $338.29
Only null left
 
18607853
View Documents
Spectrum Chemical Mfg Cor
Z107750KGBL
50 kg Fiber Drum
Each for $7,782.14
Only null left
 
Description

Description

Spectrum™ Chemical Zinc Gluconate, USP is used for treatment and prevention of zinc deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

4468-02-4
455.67
WHMDKBIGKVEYHS-IYEMJOQQSA-L
[Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
C12H22O14Zn
MFCD00868110
zinc(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)

Specifications

4468-02-4
5.5 to 7.5
C12H22O14Zn
500 g
[Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
455.67
USP
1
Poly Bottle
MFCD00868110
WHMDKBIGKVEYHS-IYEMJOQQSA-L
zinc(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
97 to 102%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'