missing translation for 'onlineSavingsMsg'
Learn More

Zinc foil, 0.25mm (0.01in) thick, 30cm (12in) wide, 99.98% (metals basis)

Form: approx. 53g/30x10cm | CAS: 7440-66-6 | Zn | 65.38 g/mol

$86.67 - $600.21

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10436BS
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
010436BS
10 cm
Each for $86.67
Only null left
 
AA10436BW
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
010436BW
50 cm
Each for $136.64
Only null left
 
AA10436CB
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
010436CB
250 cm
Each for $600.21
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]

Specifications

Gray
Gray foil
Odorless
Foil
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
Zinc
7440-66-6
10 cm
99.98% (Metals basis)
Zn
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
65.38
CHEBI:30185
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-175-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only