missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

$3447.00 - $13296.33

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
02004588
View Documents
Avantor J.T.Baker
429601
500 g Poly Bottle
Case of 4 Each for $3,447.00
Only null left
 
02004587
View Documents
Avantor
429605
2.5 kg Poly Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

5970-45-6
219.498
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
zinc;diacetate;dihydrate
C4H10O6Zn
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
2724192
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

5970-45-6
500 g
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
219.498
219.51
Poly Bottle
Zinc Acetate, Dihydrate
Crystals
C4H10O6Zn
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc;diacetate;dihydrate
2724192
BAKER ANALYZED™ ACS Reagent
1L = 1.74kg
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'