missing translation for 'onlineSavingsMsg'
Learn More

ZFP36 Monoclonal Antibody (K94030_9A2), Invitrogen™

Mouse Monoclonal Antibody

$1557.73 - $2745.56

Specifications

Antigen ZFP36
Clone K94030_9A2
Concentration 1 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA555704
View Documents
Invitrogen™
MA555704
50 μg
Each for $1,557.73
Only null left
 
MA556084
View Documents
Invitrogen™
MA556084
200 μg
Each for $2,745.56
Only null left
 
Description

Description

Sequence of this protein is as follows: DLAAPGHPPV LRQSISFSGL PSGRRTSPPP PGLAGPSLSS SSFSPSSSPP PPGDLPLSPS AFSAAPGTPL ARRDPTPVCC PSCRRATPIS VWGPLGGLVR TPSVQSLGSD PDEYASSGSS LGGSDSPVFE AGVFAPPQPV

ZFP36 Monoclonal antibody specifically detects ZFP36 in Human samples. It is validated for Immunoprecipitation, Western Blot
Specifications

Specifications

ZFP36
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide
G0/G1 switch regulatory protein 24; G0S24; GOS24; growth factor-inducible nuclear protein NUP475; mRNA decay activator protein ZFP36; Nup475; RNF162A; Tax interacting protein; TIS11; TIS11A; TIS11D; TISII; TNF alpha inhibitor; TPA-induced sequence 11; Tristetraprolin; tristetraprolin; LOW QUALITY PROTEIN: mRNA decay activator protein ZFP36; tristetraproline; TTP; Zfp36; zfp-36; ZFP36 ring finger protein; zinc finger protein 36; zinc finger protein 36 homolog; zinc finger protein 36, C3H type, homolog; zinc finger protein, C3H type, 36 homolog
Recombinant human ZFP36 (immunogen range is 171-310).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Zfp36
K94030_9A2
Immunoprecipitation, Western Blot
Unconjugated
Mouse
Human
P26651
7538
IgG2a
Protein A
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'