Learn More
YSI™ 6051020 Pro Series pH/ORP/ISE and DO Field Cable
For use with Pro Plus and Pro1020 instruments.
Supplier:  YSI™ 605102030
Catalog No. 15-177-485
Only null left
Explore available promotions
Description
Description
  • 2-port cable
  • One sensor port accepts a pH, ORP, or ISE sensor while the second port accepts a dissolved oxygen (DO) sensor
  • Cable Length: 30m
  • Includes a cable management kit and integrated temperature sensor
Specifications
Specifications
Pro Plus and Pro1020 Instruments
Cable management kit and integrated temperature sensor
30 m
Product Suggestions
Product Suggestions