missing translation for 'onlineSavingsMsg'
Learn More

Xanthydrol for the Detection of Urea, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

$412.31 - $1347.41

Chemical Identifiers

MDL Number MFCD00005057
Synonym 9-Hydroxyxanthene; 9-Xanthenol
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111015864
View Documents
Merck Emd Millipore
9558025G
25 g
Each for $412.31
Only null left
 
111015865
View Documents
Merck Emd Millipore
95580100G
100 g
Each for $1,347.41
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00005057
9-Hydroxyxanthene; 9-Xanthenol

Specifications

123°C to 128°C; 127°C to 128°C
C13H10O2
25 g
198.22
90-46-0
MFCD00005057
9-Hydroxyxanthene; 9-Xanthenol
≥99.0% (HPLC)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'