missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xanthosine dihydrate, 99%

CAS: 5968-90-1 | C10H12N4O6·2H2O | 320.26 g/mol

$107.98 - $346.79

Chemical Identifiers

CAS 5968-90-1
Molecular Formula C10H12N4O6·2H2O
Molecular Weight (g/mol) 320.26
InChI Key AQQAMLPKPXTPOL-GWTDSMLYSA-N
Synonym xanthosine dihydrate, unii-1u62i480sp, 9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purine-2,6-diol dihydrate, xanthosine, dihydrate, xanthosine dihydrate mi, xanthosine,dihydrate, 9-beta-d-ribofuranosyl xanthine dihydrate, 9-, a-d-ribofuranosylxanthine, xanthosine, dihydrate 8ci,9ci
PubChem CID 91886582
IUPAC Name 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purine-2,6-dione;hydrate
SMILES C1=NC2=C(N1C3C(C(C(O3)CO)O)O)NC(=O)NC2=O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC207630010
View Documents
Thermo Scientific Chemicals
207630010
1 g
Each for $107.98
Only null left
 
AC207630050
SDS View Documents
Thermo Scientific Chemicals
207630050
5 g
Each for $346.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5968-90-1
320.26
xanthosine dihydrate, unii-1u62i480sp, 9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purine-2,6-diol dihydrate, xanthosine, dihydrate, xanthosine dihydrate mi, xanthosine,dihydrate, 9-beta-d-ribofuranosyl xanthine dihydrate, 9-, a-d-ribofuranosylxanthine, xanthosine, dihydrate 8ci,9ci
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purine-2,6-dione;hydrate
C10H12N4O6·2H2O
AQQAMLPKPXTPOL-GWTDSMLYSA-N
91886582
C1=NC2=C(N1C3C(C(C(O3)CO)O)O)NC(=O)NC2=O.O

Specifications

5968-90-1
98.5% min. (HPLC)
C10H12N4O6·2H2O
14,10065
-53
AQQAMLPKPXTPOL-GWTDSMLYSA-N
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purine-2,6-dione;hydrate
91886582
99%
Authentic
Glass Bottle
1 g
−53.00 (22.00°C c=8, 0.3N NaOH)
xanthosine dihydrate, unii-1u62i480sp, 9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purine-2,6-diol dihydrate, xanthosine, dihydrate, xanthosine dihydrate mi, xanthosine,dihydrate, 9-beta-d-ribofuranosyl xanthine dihydrate, 9-, a-d-ribofuranosylxanthine, xanthosine, dihydrate 8ci,9ci
C1=NC2=C(N1C3C(C(C(O3)CO)O)O)NC(=O)NC2=O.O
320.26
320.26
Xanthosine dihydrate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only