missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Wood Handled Cotton Swabs and Applicators
Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens, and for cleaning delicate instrument.
Supplier: Fisherbrand™ 22363168
Description
- Multi-purpose applicators ideal for specimen collection and use in a wide range of diagnostic testing and screening
- Sterile and non-sterile products available (see product specifications)
- Polyester-tipped and cotton applicators on wooden or plastic handles available (see product specifications)
- Not suitable for PCR DNA/ RNA testing
Specifications
6 in. | |
Small tip cotton swab | |
Cotton | |
Autoclavable | |
Non-sterile |
Wood | |
Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens and for cleaning delicate instruments | |
1000/pk | |
15 cm |