Learn More
Bulwark Women's FR/CP Lab Coat
Flame resistant/chemical protective lab coat brings together the latest innovations in lab safety to protect wearers against multiple hazards.
$319.75 - $319.75
Specifications
Gender Women's
Material Aramid Fiber
Color Royal Blue
Closure Type Snaps
Cuff Style Knit
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-999-540
View Documents
Bulwark
KNR7RBXS0R
X-Small
Each for $319.75
Only null left
19-999-541
View Documents
Bulwark
KNR7RBS0R
Small
Each for $319.75
Only null left
19-999-542
View Documents
Bulwark
KNR7RBM0R
Medium
Each for $319.75
Only null left
19-999-543
View Documents
Bulwark
KNR7RBL0R
Large
Each for $319.75
Only null left
19-999-544
View Documents
Bulwark
KNR7RBXL0R
X-Large
Each for $319.75
Only null left
19-999-545
View Documents
Bulwark
KNR7RB2X0R
2X-Large
Each for $319.75
Only null left
Description
Description
Flame resistant/chemical protective lab coat brings together the latest innovations in lab safety to protect wearers against multiple hazards.
  • Lightweight and breathable fire resistant/chemical protective lab coat.
  • Black lay-down collar for easy recognition as a CP lab coat.
  • Splash resistant pass-through for inner pocket access.
  • Flame-resistant 4.5 oz. Nomex IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame.
  • Repels small amounts of liquid chemicals that are either dropped or splashed on the fabric, resisting penetration and wicking of an inadvertent, secondary liquid chemical splash.
  • Concealed snap front neck closures.
  • One double-stitched chest pocket and two double-stitched lower pockets with flaps
  • Arc Rating ATPV 5.6 cal/cm2
  • Category 1 Protection
  • NFPA 2112 Compliant
Specifications
Specifications
Women's
Royal Blue
Knit
Non-sterile
Regular
Lay-down
Aramid Fiber
Snaps
Reusable
Flame, Chemicals
3
Category 1, NFPA 2112
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'